- Positioning of cysteine residues within the N-terminal portion of the BST-2/tetherin ectodomain is important for functional dimerization of BST-2.
Positioning of cysteine residues within the N-terminal portion of the BST-2/tetherin ectodomain is important for functional dimerization of BST-2.
BST-2/tetherin is a cellular host factor capable of restricting the release of a variety of enveloped viruses, including HIV-1. Structurally, BST-2 consists of an N-terminal cytoplasmic domain, a transmembrane domain, an ectodomain, and a C-terminal membrane anchor. The BST-2 ectodomain encodes three cysteine residues in its N-terminal half, each of which can contribute to the formation of cysteine-linked dimers. We previously reported that any one of the three cysteine residues is sufficient to produce functional BST-2 dimers. Here we investigated the importance of cysteine positioning on the ectodomain for functional dimerization of BST-2. Starting with a cysteine-free monomeric form of BST-2, individual cysteine residues were reintroduced at various locations throughout the ectodomain. The resulting BST-2 variants were tested for expression, dimerization, surface presentation, and inhibition of HIV-1 virus release. We found significant flexibility in the positioning of cysteine residues, although the propensity to form cysteine-linked dimers generally decreased with increasing distance from the N terminus. Interestingly, all BST-2 variants, including the one lacking all three ectodomain cysteines, retained the ability to form non-covalent dimers, and all of the BST-2 variants were efficiently expressed at the cell surface. Importantly, not all BST-2 variants capable of forming cysteine-linked dimers were functional, suggesting that cysteine-linked dimerization of BST-2 is necessary but not sufficient for inhibiting virus release. Our results expose new structural constraints governing the functional dimerization of BST-2, a property essential to its role as a restriction factor tethering viruses to the host cell.