Merck
  • Solubility and some crystallization properties of conglomerate forming chiral drug guaifenesin in water.
Solubility and some crystallization properties of conglomerate forming chiral drug guaifenesin in water.
Journal of pharmaceutical sciences (2014-08-06)
Robert R Fayzullin, Heike Lorenz, Zemfira A Bredikhina, Alexander A Bredikhin, Andreas Seidel-Morgenstern
ABSTRACT
The solubility of 3-(2-methoxyphenoxy)-propane-1,2-diol, the well-known chiral drug guaifenesin 1, in water has been investigated by means of polythermal and isothermal approaches. It was found that the solubilities of racemic and enantiomeric diols rac- and (R)-1 depend strongly on temperature. The ternary phase diagram of the guaifenesin enantiomers in water in the temperature range between 10°C and 40°C was constructed. Clear evidence was obtained that rac-1 crystallizes as a stable conglomerate. The Meyerhoffer coefficient for the guaifenesin-water system is more than two and strongly depends on temperature. Neither crystalline hydrates nor polymorphs were detected within the range of conditions covered. Metastable zone width data with regard to primary nucleation were also collected for rac-1 and (R)-1. On the basis of the knowledge acquired, the resolution of racemic guaifenesin by preferential crystallization from solution could be realized successfully.
Product Description
Supelco
Guaifenesin, certified reference material, pharmaceutical secondary standard
Pricing and availability is not currently available.
Guaifenesin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Guaifenesin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.