Merck
  • CCR1 and CC chemokine ligand 5 interactions exacerbate innate immune responses during sepsis.
CCR1 and CC chemokine ligand 5 interactions exacerbate innate immune responses during sepsis.
Journal of immunology (Baltimore, Md. : 1950) (2004-11-24)
Traci L Ness, Kristin J Carpenter, Jillian L Ewing, Craig J Gerard, Cory M Hogaboam, Steven L Kunkel
ABSTRACT
CCR1 has previously been shown to play important roles in leukocyte trafficking, pathogen clearance, and the type 1/type 2 cytokine balance, although very little is known about its role in the host response during sepsis. In a cecal ligation and puncture model of septic peritonitis, CCR1-deficient (CCR1(-/-)) mice were significantly protected from the lethal effects of sepsis when compared with wild-type (WT) controls. The peritoneal and systemic cytokine profile in CCR1(-/-) mice was characterized by a robust, but short-lived and regulated antibacterial response. CCR1 expression was not required for leukocyte recruitment, suggesting critical differences extant in the activation of WT and CCR1(-/-) resident or recruited peritoneal cells during sepsis. Peritoneal macrophages isolated from naive CCR1(-/-) mice clearly demonstrated enhanced cytokine/chemokine generation and antibacterial responses compared with similarly treated WT macrophages. CCR1 and CCL5 interactions markedly altered the inflammatory response in vivo and in vitro. Administration of CCL5 increased sepsis-induced lethality in WT mice, whereas neutralization of CCL5 improved survival. CCL5 acted in a CCR1-dependent manner to augment production of IFN-gamma and MIP-2 to damaging levels. These data illustrate that the interaction between CCR1 and CCL5 modulates the innate immune response during sepsis, and both represent potential targets for therapeutic intervention.
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)