Merck
  • Structure-activity relationship, biological, and pharmacological characterization of the proline sulfonamide ACT-462206: a potent, brain-penetrant dual orexin 1/orexin 2 receptor antagonist.
Structure-activity relationship, biological, and pharmacological characterization of the proline sulfonamide ACT-462206: a potent, brain-penetrant dual orexin 1/orexin 2 receptor antagonist.
ChemMedChem (2014-08-26)
Christoph Boss, Catherine Roch-Brisbare, Michel A Steiner, Alexander Treiber, Hendrik Dietrich, Francois Jenck, Markus von Raumer, Thierry Sifferlen, Christine Brotschi, Bibia Heidmann, Jodi T Williams, Hamed Aissaoui, Romain Siegrist, John Gatfield
ABSTRACT
The orexin system consists of two G-protein-coupled receptors, the orexin 1 and orexin 2 receptors, widely expressed in diverse regions of the brain, and two peptide agonists, orexin A and orexin B, which are produced in a small assembly of neurons in the lateral hypothalamus. The orexin system plays an important role in the maintenance of wakefulness. Several compounds (almorexant, SB-649868, suvorexant) have been in advanced clinical trials for treating primary insomnia. ACT-462206 is a new, potent, and selective dual orexin receptor antagonist (DORA) that inhibits the stimulating effects of the orexin peptides at both the orexin 1 and 2 receptors. It decreases wakefulness and increases non-rapid eye movement (non-REM) and REM sleep while maintaining natural sleep architectures in rat and dog electroencephalography/electromyography (EEG/EMG) experiments. ACT-462206 shows anxiolytic-like properties in rats without affecting cognition and motor function. It is therefore a potential candidate for the treatment of insomnia.
Sigma-Aldrich
ACT-462206, ≥98% (HPLC)