Merck
  • Design of efficient thermally activated delayed fluorescence materials for pure blue organic light emitting diodes.
Design of efficient thermally activated delayed fluorescence materials for pure blue organic light emitting diodes.
Journal of the American Chemical Society (2012-08-31)
Qisheng Zhang, Jie Li, Katsuyuki Shizu, Shuping Huang, Shuzo Hirata, Hiroshi Miyazaki, Chihaya Adachi
ABSTRACT
Efficient thermally activated delayed fluorescence (TADF) has been characterized for a carbazole/sulfone derivative in both solutions and doped films. A pure blue organic light emitting diode (OLED) based on this compound demonstrates a very high external quantum efficiency (EQE) of nearly 10% at low current density. Because TADF only occurs in a bipolar system where donor and acceptor centered (3)ππ* states are close to or higher than the triplet intramolecular charge transfer ((3)CT) state, control of the π-conjugation length of both donor and acceptor is considered to be as important as breaking the π-conjugation between them in blue TADF material design.
Sigma-Aldrich
3,6-Di-tert-butylcarbazole, 97%
Sigma-Aldrich
DPEPO