Merck
  • Tetrabenzyl pyrophosphate as a new class of neutral carrier responsive to lead ion.
Tetrabenzyl pyrophosphate as a new class of neutral carrier responsive to lead ion.
Talanta (2008-10-31)
D Xu, T Katsu
ABSTRACT
Tetrabenzyl pyrophosphate and diphenylphosphinic anhydride, with two phosphoryl groups (PO) as ligating sites, can be used as novel ionophores to make Pb(2+)-selective membrane electrodes. A good result was obtained with tetrabenzyl pyrophosphate, and the electrode based on this ionophore and bis(1-butylpentyl) adipate as a solvent mediator in a poly(vinyl chloride) membrane matrix exhibited a near-Nernstian response to Pb(2+) in the concentration range of 1x10(-5)-1x10(-2) M with a slope of 28.7 mV per concentration decade in a solution containing 0.1 M Mg(NO(3))(2). The limit of detection was 3x10(-6) M. The selectivity of this electrode to other metal cations was comparable to the best case in many Pb(2+)-selective electrodes so far developed. Addition of potassium tetrakis(p-chlorophenyl)borate (40 mol% relative to tetrabenzyl pyrophosphate) caused a drastic change in the response slope (53.3 mV per concentration decade), probably due to the formation of PbA(+), where A stands for anions present in the sample solution, and decreased significantly the electrode selectivity to other metal cations.
Sigma-Aldrich
Tetrabenzyl pyrophosphate, 98%
Pricing and availability is not currently available.