Merck
  • Gomisin A inhibits lipopolysaccharide-induced inflammatory responses in N9 microglia via blocking the NF-κB/MAPKs pathway.
Gomisin A inhibits lipopolysaccharide-induced inflammatory responses in N9 microglia via blocking the NF-κB/MAPKs pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2013-11-12)
Xiaoxiao Wang, Di Hu, Lijia Zhang, Guoning Lian, Siqi Zhao, Chunming Wang, Jun Yin, Chunfu Wu, Jingyu Yang
ABSTRACT
Gomisin A, one of the major dibenzocyclooctadiene lignans isolated from Schisandra chinensis Baill., has proved to possess a variety of pharmacological effects. The aim of the present study was to investigate the anti-inflammatory and neuroprotective effects of gomisin A as well as its potential molecular mechanisms. It was found that gomisin A not only inhibited the production of NO and PGE2 in a concentration-dependent manner but also suppressed the expressions of iNOS and COX-2 in LPS-stimulated N9 microglia without observable cytotoxicity. Gomisin A was also able to attenuate the mRNA expression and the production of pro-inflammatory factors TNF-α, IL-1β and IL-6. Moreover, LPS induced reactive oxygen species (ROS) production, NADPH oxidase activation, and gp91phox expression, which were markedly inhibited by gomisin A in microglia. Furthermore, the data showed that gomisin A significantly down-regulated the TLR4 protein expression, and inhibited nuclear transcription factor (NF)-κB and mitogen-activated protein kinases (MAPKs) signaling pathways. Additionally, gomisin A alleviated the cell death of SH-SY5Y neuroblastoma, rat primary cortical and hippocampal neurons induced by the conditioned-media from activated microglia. In summary, gomisin A may exert neuroprotective effects by attenuating the microglia-mediated neuroinflammatory response via inhibiting the TLR4-mediated NF-κB and MAPKs signaling pathways.
Sigma-Aldrich
Gomisin A