Merck
  • [Antiradical activity of 3-substituted coumarins and their effect on iron-dependent chemiluminescence].
[Antiradical activity of 3-substituted coumarins and their effect on iron-dependent chemiluminescence].
Biulleten' eksperimental'noi biologii i meditsiny (1991-10-01)
Iu A Vladimirov, E A Parfenov, O M Epanchintseva, L N Smirnov
ABSTRACT
The antioxidant capacity of 3-aminocoumarin, 3-oxycoumarin, 3-acetylaminocoumarin, and 3-coumarin carbonic acid has been investigated with chemiluminescence measurement and by the accumulation of TBA-active products. All coumarins were found to be antioxidants, with 3-oxy-, 3-amino- and 3-acetylamino coumarins being capable of amplifying chemiluminescence at early stages of the process.
Sigma-Aldrich
3-Aminocoumarin, 97%