- Mouse alpha N-catenin: two isoforms, specific expression in the nervous system, and chromosomal localization of the gene.
Mouse alpha N-catenin: two isoforms, specific expression in the nervous system, and chromosomal localization of the gene.
We isolated cDNAs encoding mouse homologues of chicken alpha N-catenin, a protein associated with the cadherin cell adhesion molecules, and identified two isoforms of this protein. One isoform (alpha N-catenin I) was identical to the chicken alpha N-catenin that had previously been identified, and the other (alpha N-catenin II) differed in having a 48-amino acid insertion in its C-terminal region. The ratio of the two isoforms changed during development; the isoform II was more abundant than the other in earlier embryonic stages, whereas isoform I was predominant in the adult stage. Immunostaining and in situ hybridization analyses revealed that the mouse alpha N-catenin was expressed almost exclusively in the nervous system. During embryogenesis, alpha N-catenin was first detected in nerve fibers of cranial and dorsal root ganglia and also in early neurons in the neural tube, including motor neurons. Thereafter, the expression of this protein occurred in various regions of the nervous system. Neurons, in general, strongly expressed alpha N-catenin, especially in their axonal fibers. On the other hand, the expression in glial cells varied with the region. For example, the ependymal layers of the neural tube generally expressed low levels of alpha N-catenin except at the inner limiting membrane facing the central canal, whereas the floor and roof plate exhibited strong expression of this protein at various portions of the central nervous system. The choroid plexus was devoid of alpha N-catenin. In the alpha N-catenin-negative regions, another subtype of alpha-catenin, alpha E-catenin, was expressed. Concerning nonneural tissues, alpha N-catenin was expressed only in some local mesenchymal cell clusters and the lens fibers. These results suggest that alpha N-catenin plays specific roles in neural cell-cell interactions. We also localized the mouse alpha N-catenin gene to chromosome 6.