Merck
  • Photochemical Reactions of Fluorinated Pyridines at Half-Sandwich Rhodium Complexes: Competing Pathways of Reaction.
Photochemical Reactions of Fluorinated Pyridines at Half-Sandwich Rhodium Complexes: Competing Pathways of Reaction.
Organometallics (2014-02-25)
Barbara Procacci, Robin J Blagg, Robin N Perutz, Nuria Rendón, Adrian C Whitwood
ABSTRACT
Irradiation of CpRh(PMe
Sigma-Aldrich
Pentafluoropyridine, ≥99%