Merck
  • Rigid NON- and NSN-ligand complexes of tetravalent and trivalent uranium: comparison of U-OAr2 and U-SAr2 bonding.
Rigid NON- and NSN-ligand complexes of tetravalent and trivalent uranium: comparison of U-OAr2 and U-SAr2 bonding.
Dalton transactions (Cambridge, England : 2003) (2012-05-09)
Balamurugan Vidjayacoumar, Sougandi Ilango, Matthew J Ray, Terry Chu, Kristopher B Kolpin, Nicholas R Andreychuk, Carlos A Cruz, David J H Emslie, Hilary A Jenkins, James F Britten
ABSTRACT
A rigid NSN-donor proligand, 4,5-bis(2,6-diisopropylanilino)-2,7-di-tert-butyl-9,9-dimethylthioxanthene (H(2)[TXA(2)], 1) was prepared by palladium-catalyzed coupling of 2,6-diisopropylaniline with 4,5-dibromo-2,7-di-tert-butyl-9,9-dimethylthioxanthene. Deprotonation of 1 using (n)BuLi provided Li(2)(DME)(2)[TXA(2)] (2), and subsequent reaction with UCl(4) afforded [Li(DME)(3)][(TXA(2))UCl(3)] (4). The analogous NON-donor ligated complex [(XA(2))UCl(3)K(DME)(3)] [3; XA(2) = 4,5-bis(2,6-diisopropylanilino)-2,7-di-tert-butyl-9,9-dimethylxanthene] was prepared by the reaction of K(2)(DME)(x)[XA(2)] with UCl(4). A cyclic voltammogram (CV) of 3 in THF/[NBu(4)][B(C(6)F(5))(4)] at 200 mV s(-1) showed an irreversible reduction to uranium(III) at E(pc) = -2.46 V versus FeCp(2)(0/+1), followed by a product wave at E(1/2) = -1.83 V. Complex 4 also underwent irreversible reduction to uranium(iii) [E(pc) = -2.56 V], resulting in an irreversible product peak at E(pa) = -1.83 V. One-electron reduction of complexes 3 and 4 using K(naphthalenide) under an argon atmosphere in DME yielded 6-coordinate [(XA(2))UCl(DME)] (5) and the thermally unstable 7-coordinate [(TXA(2))U(DME)Cl(2)Li(DME)(2)] (6), respectively. The U-S distances in 4 and 6 are uncommonly short, the C-S-U angles are unusually acute, and the thioxanthene backbone of the TXA(2) ligand is significantly bent. By contrast, the xanthene backbone in XA(2) complexes 3 and 5 is planar. However, κ(3)-coordination and an approximately meridional arrangement of the ancillary ligand donor atoms is maintained in all complexes. DFT and Atoms in Molecules (AIM) calculations were carried out on 3, 4, 5, 6, [(XA(2))UCl(3)](-) (3B), [(TXA(2))UCl(2)(DME)](-) (6B) and [(TXA(2))UCl(DME)] (6C) to probe the extent of covalency in U-SAr(2) bonding relative to U-OAr(2) bonding.
Sigma-Aldrich
2,6-Diisopropylaniline, 97%
Sigma-Aldrich
2,6-Diisopropylaniline, technical grade, 90%