Merck
  • Synthesis and antiviral evaluation of α-L-2'-deoxythreofuranosyl nucleosides.
Synthesis and antiviral evaluation of α-L-2'-deoxythreofuranosyl nucleosides.
European journal of medicinal chemistry (2011-06-15)
Kiran S Toti, Marco Derudas, Christopher McGuigan, Jan Balzarini, Serge Van Calenbergh
ABSTRACT
The synthesis of a series of α-L-2'-deoxythreofuranosyl nucleosides featuring the nucleobases A, T, C and U is described in seven steps from 1,2-O-isopropyledene-α-L-threose, involving a Vorbrüggen coupling and a Barton-McCombie deoxygenation protocol as the key steps. All analogues, including a phosphoramidate nucleoside phosphate prodrug of the T analogue, were evaluated against a broad panel of different viruses but found inactive, while also lacking notable cellular toxicity. The thymidine analogue showed inhibition to mitochondrial thymidine kinase-2 (TK-2), herpes simplex virus type 1 (HSV-1) TK, varicella-zoster virus (VZV) TK and Mycobacterium tuberculosis thymidylate kinase.
Sigma-Aldrich
O-(p-Tolyl) chlorothionoformate, ≥97.0%