Merck
  • Ampicillin concentrations in human serum and periodontal membrane following a single oral administration of talampicillin.
Ampicillin concentrations in human serum and periodontal membrane following a single oral administration of talampicillin.
General pharmacology (1988-01-01)
Y Akimoto, A Uda, H Omata, H Nishimura, M Komiya, K Kaneko, A Fujii, T Tamura
ABSTRACT
1. Ampicillin concentrations in human serum and periodontal membrane after a single oral administration of talampicillin (500 mg) were assayed by the agar diffusion (paper disc) method. 2. The peak times of serum and periodontal membrane were identical, being 150 min after administration. 3. The peak concentrations of serum and periodontal membrane were 7.81 micrograms/ml and 4.11 micrograms/g, respectively. 4. The mean ratio of periodontal membrane to serum concentration at the peak time was 0.53.
Product Description
Talampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard