Merck
Otomycosis: Diagnosis and treatment.
Clinics in dermatology (2010-03-30)
Irina Vennewald, Eckart Klemm
ABSTRACT
Aspergillus and Candida spp are the most frequently isolated fungi in patients with otomycosis. The diagnosis of otitis externa relies on the patient's history, otoscopic examination under microscopic control, and imaging studies. Direct preparation of the specimens, particularly with optical brighteners, mycologic culture, and histologic examination, is very important and strongly recommended for the correct diagnosis. Patients with noninvasive fungal otitis externa should be treated with intense débridement and cleansing, and topical antifungals. Topical antifungals, such as clotrimazole, miconazole, bifonazole, ciclopiroxolamine, and tolnaftate, are potentially safe choices for the treatment of otomycosis, especially in patients with a perforated eardrum. The oral triazole drugs, itraconazole, voriconazole, and posaconazole are effective against Candida and Aspergillus, with good penetration of bone and the central nervous system. These drugs are essential in the treatment of patients with malignant fungal otitis externa complicated by mastoiditis and meningitis.
Product Description
Isoconazole nitrate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Isoconazole, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.