Merck
  • Combination of enzyme- and Lewis acid-catalyzed reactions: a new method for the synthesis of 6,7-dihydrobenzofuran-4(5H)-ones starting from 2,5-dimethylfuran and 1,3-cyclohexanediones.
Combination of enzyme- and Lewis acid-catalyzed reactions: a new method for the synthesis of 6,7-dihydrobenzofuran-4(5H)-ones starting from 2,5-dimethylfuran and 1,3-cyclohexanediones.
Organic & biomolecular chemistry (2013-07-26)
Chimène Asta, Dietmar Schmidt, Jürgen Conrad, Wolfgang Frey, Uwe Beifuss
ABSTRACT
The Lewis acid-catalyzed domino 1,2-addition/1,4-addition/elimination between (Z)-3-hexene-2,5-dione and 1,3-dicarbonyls delivers 3-methyl-6,7-dihydrobenzofuran-4(5H)-ones exclusively with yields up to 82%. The combination of this new process with the laccase-catalyzed formation of (Z)-3-hexene-2,5-dione by oxidative cleavage of 2,5-dimethylfuran allows for the synthesis of 6,7-dihydrobenzofuran-4(5H)-ones starting directly from 2,5-dimethylfuran.
Sigma-Aldrich
1,3-Cyclohexanedione, 97%
Sigma-Aldrich
Laccase from Trametes versicolor, powder, light brown, ≥0.5 U/mg
Sigma-Aldrich
Laccase from Agaricus bisporus, powder, deep brown, ≥4 U/mg