Merck
  • Palladium-Catalyzed Carbonylation and Coupling Reactions of Aryl Chlorides and Amines.
Palladium-Catalyzed Carbonylation and Coupling Reactions of Aryl Chlorides and Amines.
The Journal of organic chemistry (1996-10-18)
Robert J. Perry, B. David Wilson
ABSTRACT
The palladium-catalyzed amidation of electron-deficient aryl chlorides proceeds readily in the presence of low CO pressures and a slight excess of an iodide salt. The rates of amidation are accelerated over those without added salt, and iodide is preferred over bromide or chloride. More electron-rich aryl chlorides were not effectively amidated, either with or without added iodide. We postulate that an intermediate anionic palladium(0) iodide complex is responsible for the enhanced reactivity.
Sigma-Aldrich
4-Chlorophenyl phenyl sulfone, ≥97%