Merck
  • Asymmetric synthesis of hexapropionate synthons by sequential enantiotopic group selective enolization of meso diketones.
Asymmetric synthesis of hexapropionate synthons by sequential enantiotopic group selective enolization of meso diketones.
Organic letters (2006-06-02)
Dale E Ward, H Martin Gillis, Olukayode T Akinnusi, M Abdul Rasheed, K Saravanan, Pradip K Sasmal
ABSTRACT
Meso 1,9-diketones (six to seven stereocenters) are readily obtained by stepwise or simultaneous two-directional aldol reactions of tetrahydro-4H-thiopyran-4-one with a thiopyran-derived aldehyde or dialdehyde. Enantioselective enolizations of these diketones with the lithium amide from (R,R)-bis(1-phenylethyl)amine occur with simultaneous kinetic resolution to give the mono-TMS enol ethers in >90% yields (BORSM) and >95% ee. The products are applicable to polypropionate synthesis. [reaction: see text]
Sigma-Aldrich
Tetrahydro-4H-thiopyran-4-one, 99%