Merck
Electrokinetics of polar liquids in contact with nonpolar surfaces.
Langmuir : the ACS journal of surfaces and colloids (2013-05-25)
Chih-Hsiu Lin, Gregory S Ferguson, Manoj K Chaudhury
ABSTRACT
Zeta potentials of several polar protic (water, ethylene glycol, and formamide) as well as polar aprotic (dimethyl sulfoxide) liquids were measured in contact with three nonpolar surfaces using closed-cell electroosmosis. The test surfaces were chemisorbed monolayers of alkyl siloxanes, fluoroalkyl siloxanes, and polydimethylsiloxanes (PDMS) grafted on glass slides. All these liquids exhibited substantial electrokinetics in contact with the nonpolar surfaces with these observations: the electrokinetic effect on the fluorocarbon-coated surface is the strongest and on a PDMS grafted surface, the effect is the weakest. Even though these hygroscopic liquids contain small amounts of water, the current models of charging based on the adsorption of hydroxide ions at the interface or the dissociation of pre-existing functionalities (e.g., silanol groups) appear to be insufficient to account for the various facets of the experimental observations. The results illustrate how ubiquitous the phenomenon of electrokinetics is with polar liquids contacting such apparently passive nonpolar surfaces. We hope that these results will inspire further experimental and theoretical studies in this important area of research that has potential practical implications.
Sigma-Aldrich
Mixed Bed Resin, DNA grade, BioReagent