Merck
  • The degradation of 1-phenylalkanes by an oil-degrading strain of Acinetobacter lwoffi.
The degradation of 1-phenylalkanes by an oil-degrading strain of Acinetobacter lwoffi.
Antonie van Leeuwenhoek (1985-01-01)
O O Amund, I J Higgins
ABSTRACT
An oil-degrading bacterium identified as Acinetobacter lwoffi was isolated by elective culture on North Sea Forties crude oil from an activated sludge sample. It grew on a wide range of n-alkanes (C12-C28) and 1-phenylalkanes, including 1-phenyldodecane, 1-phenyltridecane and 1-phenyltetradecane. The organism degraded 1-phenyldodecane to phenylacetic acid which was further metabolized via homogentisic acid, whilst 1-phenyltridecane was transformed to trans-cinamic and 3-phenylpropionic acid which were not further metabolized. Evidence is presented for a relationship between aromatic amino acid catabolism and 1-phenyldodecane degradation in this organism.
Sigma-Aldrich
1-Phenyltridecane, 99%
Sigma-Aldrich
1-Phenyldodecane, 97%