Merck
  • Inactivation of mouse liver glutathione S-transferase YfYf (Pi class) by ethacrynic acid and 5,5'-dithiobis-(2-nitrobenzoic acid).
Inactivation of mouse liver glutathione S-transferase YfYf (Pi class) by ethacrynic acid and 5,5'-dithiobis-(2-nitrobenzoic acid).
The Biochemical journal (1993-08-15)
M F Phillips, T J Mantle
ABSTRACT
Mouse liver glutathione S-transferase YfYf (Pi class) reacts with [14C]ethacrynic acid to form a covalent adduct with a stoichiometry of 1 mol per mol of subunit. Proteolytic digestion of the enzyme-[14C]ethacrynic acid adduct with V8 protease produced an 11 kDa fragment containing radioactivity. Sequencing revealed this to be an N-terminal peptide (minus the first 15 residues, terminating at Glu-112) which contains only one cysteine residue (Cys-47). This is tentatively identified as the site of ethacrynic attachment. Kinetic studies reveal that glutathione S-conjugates protect against inactivation by ethacrynic acid, but the level of protection is not consistent with their potency as product inhibitors. A model is proposed in which glutathione S-conjugates and ethacrynic acid compete for the free enzyme, and a second molecule of ethacrynic acid reacts covalently with the enzyme-ethacrynic acid complex. The native protein contains one thiol reactive with 5,5'-dithiobis-(2-nitrobenzoic acid) at neutral pH. The resultant mixed disulphide, like the ethacrynic acid adduct, is inactive, but treatment with cyanide (which incorporates on a mol for mol basis) restores activity to 35% of that of the native enzyme.
Sigma-Aldrich
S-Methylglutathione
Pricing and availability is not currently available.