Merck
  • Oxidation and radical intermediates associated with the glutathione conjugation of mucochloric acid.
Oxidation and radical intermediates associated with the glutathione conjugation of mucochloric acid.
Chemical research in toxicology (1994-07-01)
R T LaLonde, S Xie, W Chamulitrat, R P Mason
ABSTRACT
The inactivation of the drinking water mutagen mucochloric acid (MCA) by reduced glutathione (GSH) was linked to the formation of an MCA-GSH conjugate, a nonmutagen in the Salmonella typhimurium (TA100) plate incorporation assay. Anaerobic formation of MCA-GSH is found now to be associated with oxidized glutathione (GSSG) and unconverted MCA. The anaerobic reaction of GSH with MCA in the presence of the radical trap 2-methyl-2-nitrosopropane (tNB; "tert-nitrosobutane") gives rise to an electron paramagnetic resonance (EPR) resulting from the overlapping spectra of two radical adducts. The first species exhibited hyperfine coupling constants of aN = 13.65 G and aH beta = 0.73 G. The second radical adduct exhibited a three-line signal of aN = 12.8 G. The first species is assigned to an adduct of the MCA radical because deuteration of MCA (5-deuterio-MCA) caused the beta-hydrogen hyperfine coupling to collapse. The second radical adduct is unaffected by the deuteration of MCA. Thus, the involvement of both GSSG and a carbon-centered MCA radical in the action of MCA on GSH is indicated.
Sigma-Aldrich
Mucochloric acid, 99%