- A new neurohypophysial peptide, seritocin ([Ser5,Ile8]-oxytocin), identified in a dryness-resistant African toad, Bufo regularis.
A new neurohypophysial peptide, seritocin ([Ser5,Ile8]-oxytocin), identified in a dryness-resistant African toad, Bufo regularis.
From the pituitary neurointermediate lobe of the African toad Bufo regularis, vasotocin, hydrin 2 (vasotocinyl-Gly) and a mesotocin-like peptide have been isolated by HPLC and characterized by mass spectrometry, amino acid sequence and chromatographic coelution with synthetic peptides. The mesotocin-like peptide has been identified as [Ser5,Ile8]-oxytocin in place of mesotocin ([Ile8]-oxytocin) found in all other amphibians investigated to date. The name seritocin is suggested. The molecule is virtually devoid of oxytocic activity on rat uterus in contrast to mesotocin. On the other hand, the molar ratio of hydrin 2 to vasotocin in the pituitary reaches 2, whereas it is about 1 in toads and frogs from temperate regions. B. regularis is an anuran species able to withstand a hot and dry season by burrowing. The possible relationship between occurrence of seritocin and adaptation to arid environment remains to be demonstrated.