Merck
  • Quantitative and qualitative effects of N10-methylfolate on high-affinity folate binding in human leukocytes.
Quantitative and qualitative effects of N10-methylfolate on high-affinity folate binding in human leukocytes.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition (1984-01-01)
J Holm, S I Hansen, J Lyngbye
ABSTRACT
N10-methylfolate acted as a potent competitive inhibitor of high-affinity [3H] folate binding in human leukocytes, while methotrexate had no effect. Furthermore, folate binding changed into a non-cooperative type in the presence of N10-methylfolate. Hence, in qualitative and quantitative respects, the substrate specificity characteristics of leukocyte folate binding resemble those of other high-affinity folate binding systems.