- Production and characterization of monoclonal antibodies to the macrocyclic trichothecene roridin A.
Production and characterization of monoclonal antibodies to the macrocyclic trichothecene roridin A.
Two murine monoclonal antibodies to the macrocyclic trichothecene roridin A are described. Screening for antibody production was performed on absorbed anti-mouse immunoglobulin serum as double-antibody solid phase, and further characterization was done on affinity-purified anti-mouse IgG serum. The antibodies, designated 5G11 and 4H10, had affinity constants for roridin A of 9.25 X 10(7) and 1.7 X 10(7) liters/mol, respectively. In monoclonal antibody-based direct enzyme immunoassays, these IgG1 antibodies had detection limits for roridin A of 0.4 ng/ml (0.02 ng per assay) and 1.8 ng/ml (0.09 ng per assay), respectively. Both antibodies were most specific for the tested macrocyclic trichothecenes. The relative cross-reactivities of antibody 5G11 with roridin A, roridin J, verrucarin A, satratoxin G, and satratoxin H were 100.0, 43.8, 16.7, 3.7, and 18.9%, respectively; for antibody 4H10 they were 100.0, 6.3, 64.0, 4.4, and 4.9%, respectively.