Merck
  • Oral contraceptive containing chlormadinone acetate and ethinylestradiol reduces plasma concentrations of matrix metalloproteinase-2 in women with polycystic ovary syndrome.
Oral contraceptive containing chlormadinone acetate and ethinylestradiol reduces plasma concentrations of matrix metalloproteinase-2 in women with polycystic ovary syndrome.
Basic & clinical pharmacology & toxicology (2012-04-19)
Valéria A Gomes, Carolina S Vieira, Anna L Jacob-Ferreira, Vanessa A Belo, Gustavo M Soares, Janaína B França, Rui A Ferriani, Jose E Tanus-Santos
ABSTRACT
Biochemical markers of cardiovascular disease, including matrix metalloproteinases (MMPs), are altered in women with polycystic ovary syndrome (PCOS), with many of these alterations thought to be due to excess androgen concentrations. Despite oral contraceptives (OCs) being the first-line pharmacological treatment in women with PCOS and the importance of MMPs in many physiological conditions and pathological states, including cardiovascular diseases, no study has yet evaluated whether OCs alter plasma concentrations of MMPs. We therefore assessed whether treatment with an OC containing the anti-androgenic progestogen alters MMP profiles in women with PCOS. We analysed 20 women with PCOS who wanted hormonal contraception (OC-PCOS group), 20 ovulatory women who required hormonal contraception (OC-control group) and 20 ovulatory women who wanted non-hormonal contraception (non-OC-control group). OC consisted of cyclic use of 2 mg chlormadinone acetate/30 μg ethinylestradiol for 6 months. Plasma concentrations of MMP-2, MMP-9, TIMP-1 and TIMP-2 were measured by gelatin zymography or enzyme-linked immunoassays. OC treatment for 6 months significantly reduced plasma MMP-2 concentrations in the OC-control and OC-PCOS groups and TIMP-2 and TIMP-1 concentrations levels in the OC-control group (all p < 0.05), but had no effects on MMP-9 concentrations or on MMP-2/TIMP-2 and MMP-9/TIMP-1 ratios in any group (all p > 0.05). These findings indicated that long-term treatment with an OC containing chlormadinone acetate plus ethinylestradiol reduced plasma MMP-2 concentrations in both healthy and PCOS women. As the latter have imbalances in circulating matrix MMPs, treatment of these women with an OC may be beneficial.
Sigma-Aldrich
Chlormadinone acetate, ≥98%