- Development of tryptase inhibitors derived from thalidomide.
Development of tryptase inhibitors derived from thalidomide.
Bioorganic & medicinal chemistry (2010-06-22)
Masashi Tetsuhashi, Minoru Ishikawa, Mariko Hashimoto, Yuichi Hashimoto, Hiroshi Aoyama
PMID20561792
ABSTRACT
A novel series of tryptase inhibitors with a N-phenylphthalimide skeleton structurally derived from thalidomide (1) has been developed. Structure-activity relationship studies led to a potent and selective tryptase inhibitor, 2-(4-cyanophenyl)isoindole-1,3-dione-5-yl 3-(2-aminopyridin-5-yl)propanoate (7), with the IC50 value of 78 nM.