Merck
  • Complexion changes in oral contraceptive users. Results from a phase IV multicenter trial evaluating the safety and efficacy of ethynodiol diacetate, 1 mg, with ethinyl estradiol, 35 micrograms.
Complexion changes in oral contraceptive users. Results from a phase IV multicenter trial evaluating the safety and efficacy of ethynodiol diacetate, 1 mg, with ethinyl estradiol, 35 micrograms.
The Journal of reproductive medicine (1991-04-01)
J M Wheeler, L R Malinak
ABSTRACT
An open-label, phase IV, multicenter survey of obstetrician-gynecologists was conducted to evaluate the efficacy and safety of a low-dose monophasic oral contraceptive, ethynodiol diacetate, 1 mg, with ethinyl estradiol, 35 micrograms. Surveys from 983 community-based physicians reported on 6,382 women. Most patients did not experience "clinically noticeable complexion changes" (5,695/6,382, or 89.2%). Of the 687 patients with complexion changes, nearly three-fourths reported an improvement (501/687, or 72.9%). A follow-up questionnaire was sent to 127 respondents (18.6%) who reported worsening of the complexion; 70% of the questionnaires were returned. Most complexion worsening was of slight degree (63%), reported by the patient and not the physician (84% vs. 16%), and experienced during the first two to three months (84%). Although the literature includes many references to skin condition "improvement" on oral contraceptives, this report of a descriptive study gives clinicians as estimate of the incidence and severity of complexion changes in actual use.