Merck
  • [Pharmacokinetics of biological tags in a case of acute valpromide self poisoning].
[Pharmacokinetics of biological tags in a case of acute valpromide self poisoning].
Annales de biologie clinique (2010-03-30)
Diane Commandeur, Marc Danguy Des Deserts, Christophe Giacardi, Sandrine Eyrieux, Mehdi Ould-Ahmed, Isabelle Drouillard
ABSTRACT
We report the case of a 35-year-old woman hospitalized in intensive care unit after valpromide self-poisoning (DĆ©pamide). The dose supposed to be taken was 15 g. The patient eleven hours after ingestion, presented, a coma with bilateral non-reactive mydriasis, justifying intubation and mechanical ventilation. An acidosis (pH = 7.24), a hyperlactatemia (2.72 mmol/L), a hyperammonemia (328.9 micromol/L), and were diagnosed valproic acid plasmatic concentration was 1190 microg/mL at 22 hours. Symptomatic treatment and L-carnitine therapy (50 mg/kg/12h) normalized the action of biological abnormalities and allowed disappearance of symptoms; the patient was extubated after 46 hours, and outcome from intensive care unit at 65 hours.
Sigma-Aldrich
Valpromide, ≥97% (NMR)