- [The effect of irradiation and alkylating agents on chromatin breakdown in normal and malignant lymphoid cells].
[The effect of irradiation and alkylating agents on chromatin breakdown in normal and malignant lymphoid cells].
Radiobiologiia (1991-01-01)
N I Riabchenko, V Jurasková, B P Ivannik, A V Konov, V Drasil
PMID2008520
ABSTRACT
Regularities of chromatin degradation in thymocytes and LS/BL tumor cells have been investigated. It has been shown that the rate of DNA degradation by Ca/Mg-dependent endonuclease in LS/BL tumor cells is 25 times lower than that in thymocytes, and radiation does not induce chromatin degradation. The alkylating agent TS 160 causes chromatin degradation in both LS/BL cells and thymocytes. In contrast to radiation TS 160 inhibits the endogenous chromatin degradation by Ca/Mg-dependent endonuclease in thymocytes.