Merck
  • Effect of casopitant, a novel NK-1 antagonist, on the pharmacokinetics of dolasetron and granisetron.
Effect of casopitant, a novel NK-1 antagonist, on the pharmacokinetics of dolasetron and granisetron.
Supportive care in cancer : official journal of the Multinational Association of Supportive Care in Cancer (2009-02-12)
Laurel M Adams, Brendan Johnson, Ke Zhang, Lin Yue, Lyndon C Kirby, Peter Lebowitz, Randall Stoltz
ABSTRACT
The objective of this study was to characterize the impact of casopitant, a novel neurokinin-1 receptor antagonist under investigation for the prevention of postoperative and chemotherapy-induced nausea and vomiting, on the pharmacokinetics of the commonly prescribed 5-hydroxytryptamine receptor 3 receptor antagonists, dolasetron or granisetron. In a phase I, open-label, two-part, two-period, single-sequence study, two cohorts of healthy subjects received either oral dolasetron (100 mg once daily for 3 days) or oral granisetron (2 mg once daily for 3 days) alone (period 1) and combined with oral casopitant, 150 mg day 1, 50 mg days 2 and 3 (period 2). Pharmacokinetics of hydrodolasetron and granisetron were assessed on days 1 and 3 of each period. Log-transformed area under the curve (AUC) and Cmax were statistically analyzed by performing an analysis of variance. Eighteen subjects were enrolled in the dolasetron cohort; nine subjects were CYP2D6 extensive metabolizers (EMs) and nine subjects were CYP2D6 poor metabolizers. Nineteen subjects were enrolled in the granisetron cohort. The largest changes in hydrodolasetron exposure after coadministration with casopitant were seen in CYP2D6 EMs, with a 24% increase in hydrodolasetron AUC on day 1 and 30% increase in Cmax on days 1 and 3. All other changes in hydrodolasetron exposure were <20%, and granisetron exposure was not altered to any relevant extent (<11%). None of the changes observed are considered clinically meaningful, and coadministration of casopitant with dolasetron or granisetron was well tolerated.
Sigma-Aldrich
Dolasetron mesylate hydrate, ≥98% (HPLC), powder