Merck
  • Photolabile protection of alcohols, phenols, and carboxylic acids with 3-hydroxy-2-naphthalenemethanol.
Photolabile protection of alcohols, phenols, and carboxylic acids with 3-hydroxy-2-naphthalenemethanol.
The Journal of organic chemistry (2008-09-11)
Anton Kulikov, Selvanathan Arumugam, Vladimir V Popik
ABSTRACT
Irradiation of alcohols, phenols, and carboxylic acids "caged" with the (3-hydroxy-2-naphthalenyl)methyl group results in fast (k(release) approximately = 10(5) s(-1)) release of the substrates with good quantum (Phi = 0.17-0.26) and chemical (>90%) yields. The initial byproduct of the photoreaction, 2-naphthoquinone-3-methide, reacts rapidly with water (k(H2O) = 144 +/- 11 s(-1)) to produce parent 3-hydroxy-2-naphthalenemethanol. The o-quinone methide intermediate can be also trapped by other nucleophiles or converted into a photostable Diels-Alder adduct with ethyl vinyl ether.
Sigma-Aldrich
2-Naphthalenemethanol, 98%
Pricing and availability is not currently available.