Merck
  • Determination of illicit drugs and their metabolites in human urine by liquid chromatography tandem mass spectrometry including relative ion intensity criterion.
Determination of illicit drugs and their metabolites in human urine by liquid chromatography tandem mass spectrometry including relative ion intensity criterion.
Journal of analytical toxicology (2007-12-21)
Marta Concheiro, Ana De Castro, Oscar Quintela, Angelines Cruz, Manuel López-Rivadulla
ABSTRACT
A method, using 0.5 mL of urine, was developed for the simultaneous determination of ecgonine methyl ester, benzoylecgonine, morphine, codeine, 6-acetylmorphine, amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxymethamphetamine (MDMA), methadone, 2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine (EDDP), and d-lysergic acid diethylamide (LSD). The analysis was performed by liquid chromatography with tandem mass spectrometry, after solid-phase extraction in the presence of their deuterated analogues. Reversed-phase separation on an Atlantis dC18 column was achieved in 12.5 min, under gradient conditions. The method was fully validated, including linearity (1-2000 microg/L for ecgonine methyl ester, benzoylecgonine, 6-acetylmorphine, methamphetamine, MDMA, and EDDP; 2-2000 microg/L for morphine, codeine, MDA, and methadone; 2-1000 microg/L for amphetamine, and 0.2-100 microg/L for LSD; r(2)>0.99); recovery (>65%), within-day and between-day precision, and accuracy (CV and MRE<15%); limit of detection (0.1 microg/L for LSD, 0.5 microg/L for ecgonine methyl ester, benzoylecgonine, methamphetamine, MDMA, 6-monoacetylmorphine, and EDDP, and 1 microg/L for amphetamine, MDA, morphine, and methadone); quantitation (lowest level of the calibration curve); relative ion intensities, freeze-and-thaw stability, and matrix effect. The procedure showed to be sensitive and specific, and was applied to real cases and quality control samples from a quality control program.