- Collecting duct carcinoma: an incidental finding in a non functional kidney secondary to nephrolithiasis.
Collecting duct carcinoma: an incidental finding in a non functional kidney secondary to nephrolithiasis.
JPMA. The Journal of the Pakistan Medical Association (2007-04-17)
Asim Qureshi
PMID17432025
ABSTRACT
A nephrectomy specimen was sent to the laboratory for end stage renal disease secondary to nephrolithiasis. Initial sections incidentally revealed a tumor infiltrating the normal renal tissue. Further workup including cytochemical and immuno-histochemical stains confirmed it to be collecting duct carcinoma.