Merck
  • Iterative deoxypropionate synthesis based on a copper-mediated directed allylic substitution: formal total synthesis of borrelidin (C3-C11 fragment).
Iterative deoxypropionate synthesis based on a copper-mediated directed allylic substitution: formal total synthesis of borrelidin (C3-C11 fragment).
Chemistry (Weinheim an der Bergstrasse, Germany) (2006-06-15)
Christian Herber, Bernhard Breit
ABSTRACT
A new iterative strategy for the flexible preparation of any oligodeoxypropionate stereoisomer is presented which relies on an o-DPPB-directed copper mediated allylic substitution employing enantiomerically pure Grignard reagents; the reaction is working with perfect control over all aspects of the reaction selectivity. This key C--C bond-forming step features reversed polarity compared with established enolate alkylation methodology. It thus avoids existing problems of enolate alkylation strategies such as enolate reactivity as well as costs and problems associated with the chiral auxiliary. Practicability of this new method is demonstrated through application in natural product syntheses. Thus, an efficient synthesis of the northern part of the angiogenesis inhibitor borrelidin (28), the deoxypropionate building block 27, could be devised, representing a formal total synthesis.
Sigma-Aldrich
Borrelidin, from Streptomyces parvulus, ā‰„98% (HPLC)