Merck
Pheromone unwrapping by pH flip-flopping.
Chemistry & biology (2004-08-25)
Walter S Leal
ABSTRACT
The Asian elephant utilizes the same sex pheromone as a number of moth species, (Z)-7-dodecen-1-yl acetate encapsulated in a serum-derived albumin. The chemical signal is emitted in the urine and received in the mucus of the trunk. The unwrapping of the package is pH mediated.