Merck
  • Effects of 5alpha-androst-16-en-3alpha-ol on the pulsatile secretion of luteinizing hormone in human females.
Effects of 5alpha-androst-16-en-3alpha-ol on the pulsatile secretion of luteinizing hormone in human females.
Chemical senses (2000-08-17)
K Shinohara, M Morofushi, T Funabashi, D Mitsushima, F Kimura
ABSTRACT
We examined the effects of 5alpha-androst-16-en-3alpha-ol (3alpha-androstenol) on pulsatile luteinizing hormone (LH) secretion in human females. The frequency of the LH pulse in the follicular phase was decreased by exposing the women to 3alpha-androstenol.
Sigma-Aldrich
5α-Androst-16-en-3α-ol