Merck
  • Glycine catalyzed convenient synthesis of 2-amino-4H-chromenes in aqueous medium under sonic condition.
Glycine catalyzed convenient synthesis of 2-amino-4H-chromenes in aqueous medium under sonic condition.
Ultrasonics sonochemistry (2012-02-14)
Bandita Datta, M A Pasha
ABSTRACT
A one-pot three-component condensation of an aldehyde, malononitrile, and resorcinol has been achieved by ultrasound method. The reaction has been catalyzed by glycine in aqueous medium. This protocol afforded corresponding 2-amino-4H-chromenes in shorter reaction times, high yields and simple work-up procedures with the green aspects by avoiding toxic reagents.
Sigma-Aldrich
Malononitrile, ≥99%
Sigma-Aldrich
Malononitrile, Arxada quality, ≥99.0% (calculated, GC, KF)