Merck
  • Obaculactone suppresses Th1 effector cell function through down-regulation of T-bet and prolongs skin graft survival in mice.
Obaculactone suppresses Th1 effector cell function through down-regulation of T-bet and prolongs skin graft survival in mice.
Biochemical pharmacology (2010-04-07)
Fangyuan Gong, Yan Shen, Qi Zhang, Yang Sun, Jiayu Tang, Feifei Tao, Qiang Xu
ABSTRACT
Allograft rejection is a predominantly Th1 immune response. In this study, we showed that obaculactone, a natural compound derived from citrus fruit, prolonged skin graft survival in mice when treated after but not before transplantation. Furthermore, obaculactone inhibited alloantigen-specific production of Th1 cytokine IFN-gamma as well as proinflammatory cytokine IL-2, TNFalpha and IL-6. In parallel, IL-10 production was markedly up-regulated. Obaculactone significantly enhanced the percentage of CD4(+)CD25(+)Foxp3(+) Treg cells in the CD4(+) splenocytes without any effect on their inhibitory function. In vitro and in vivo tests showed obaculactone down-regulated T-bet expression in Th1 effector cells. Taken together, the unique immunomodulatory properties might qualify obaculactone as a putative, therapeutic compound for the treatment of Th1-driven diseases, including transplant rejection.
Sigma-Aldrich
Limonin, from citrus seeds, >90% (HPLC)