Merck
  • High-performance liquid chromatographic method for measuring total plasma homocysteine levels.
High-performance liquid chromatographic method for measuring total plasma homocysteine levels.
Journal of chromatography. B, Biomedical sciences and applications (1998-12-30)
I Fermo, C Arcelloni, G Mazzola, A D'Angelo, R Paroni
ABSTRACT
We have modified a high-performance liquid chromatographic (HPLC) procedure based on SBD-F (ammonium-7-fluorobenzo-2-oxa-1,3-diazole-4-sulphonate) pre-column derivatization to obtain an assay that is useful for routine clinical total plasma homocysteine (tHcy) analysis. The introduction of easily handled sodium borohydride instead of the traditional tri-n-butylphosphine in dimethylformamide as a reductant and a 14-min run-time using basic isocratic HPLC equipment are the more notable advantages. The addition of mercaptopropionylglycine as an internal standard contributed to improvements in the reproducibility of the assay, yielding within- and between-run precisions of 1.9 and 4% (C.V.), respectively. Reference values for fasting tHcy were 7.65+/-2.3 and 8.9+/-2.4 micromol/l, while post-methionine load gave tHcy levels of 19.9+/-5.5 and 26.8+/-5.5 micromol/l, for women and men, respectively (n=40).
Product Description
Supelco
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, for HPLC derivatization, LiChropur, ≥98.5% (HPLC)
Sigma-Aldrich
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, ≥98%