Merck
  • Si(IV)-methoxyethylene-glycol-naphthalocyanine: synthesis and pharmacokinetic and photosensitizing properties in different tumour models.
Si(IV)-methoxyethylene-glycol-naphthalocyanine: synthesis and pharmacokinetic and photosensitizing properties in different tumour models.
Journal of photochemistry and photobiology. B, Biology (1998-02-12)
V Mantareva, M Shopova, G Spassova, D Wöhrle, S Muller, G Jori, F Ricchelli
ABSTRACT
A Si(IV)-naphthalocyanine bearing two methoxyethylenglycol axial ligands to the centrally coordinated metal ion (SiNc) was prepared by chemical synthesis and assayed for the phototherapeutic activity after administration in a Cremophor formulation to C57BI/6 mice bearing a subcutaneously transplanted Lewis lung carcinoma or B16 pigmented melanoma. Pharmacokinetic studies indicate that the maximal accumulation in the tumour occurs at 24 h after intraperitoneal injection of 0.5 mg kg-1 of SiNc, although the naphthalocyanine concentration in the Lewis lung carcinoma (0.70 microgram g-1) is significantly larger than that in the B16 pigmented melanoma (0.15 microgram g-1). This results in a higher selectivity of tumour targeting in the case of the lung carcinoma. Photodynamic therapy (782 nm, 370 mW cm-2, 360 J cm-2) at 24 h after SiNc injection causes an efficient tumour response for Lewis lung carcinoma (50% lower tumour diameter on day 19 post-treatment as compared to untreated controls) while the pigmented melanoma shows only a minor response regarding the rate of tumour growth.
Sigma-Aldrich
2,3-Naphthalocyanine, Dye content 95 %