Merck
  • The sequential nature of the negative cooperativity in rabbit muscle glyceraldehyde-3-phosphate dehydrogenase.
The sequential nature of the negative cooperativity in rabbit muscle glyceraldehyde-3-phosphate dehydrogenase.
European journal of biochemistry (1980-11-01)
Y I Henis, A Levitzki
ABSTRACT
The binding of nicotinamide--adenine dinucleotide (NAD+), nicotinamide--1,N6-ethenoadenine dinucleotide (epsilon NAD+), acetylpyridine--adenine dinucleotide (AcPyAD+), ATP, and adenosine diphosphoribose (ADP-ribose) to rabbit muscle glyceraldehyde-3-phosphate dehydrogenase (the enzyme) was examined. NAD+ and epsilon NAD+ were found to bind to the apoenzyme in a negatively cooperative manner, whereas AcPyAD+, ATP, and ADP-ribose bind non-cooperatively to the NAD+ sites. The strong negative cooperativity in coenzyme binding was found to be abolished in the presence of AcPyAD+ and strongly weakened by ATP, ADP, and AMP, but was not affected by the addition of ADP-ribose. These findings demonstrate that the mechanism of the negative cooperativity in coenzyme binding to the enzyme involves ligand-induced conformational changes between neighboring sites. These findings cannot be accounted for by the pre-existent asymmetry model. The results support our previous hypothesis that the structure of the pyridine moiety of the coenzyme analogues plays a role in orienting the adenine moiety in the adenine subsite, and thus affects the cooperativity observed in the binding of the coenzyme analogue.
Sigma-Aldrich
Nicotinamide 1,N6-ethenoadenine dinucleotide, ≥98%
Pricing and availability is not currently available.