Merck
  • Metabolism of C-terminal pentapeptide of gastrin in the rat. Part IV. Catabolic pathways of the N-terminal BOC-14C-glycine fragment.
Metabolism of C-terminal pentapeptide of gastrin in the rat. Part IV. Catabolic pathways of the N-terminal BOC-14C-glycine fragment.
Hepato-gastroenterology (1980-04-01)
L Varga, M Sasvári, B Penke, O Baláspiri, V Varró
ABSTRACT
The catabolism of the 14C-labelled pentagastrin was investigated in rats. The following results were found: -- the labelled BOC-glycine fragment is split off the pentagastrin in the small intestine -- it is absorbed through the intestinal wall, and -- enters first the portal and then the systemic circulation.
Sigma-Aldrich
Boc-Gly-OH, ≥99.0% (T)