Merck
Biotin enhances guanylate cyclase activity.
Science (New York, N.Y.) (1982-06-18)
D L Vesely
ABSTRACT
Biotin and its analog, (+)-biotin-p-nitrophenyl ester enhanced guanylate cyclase activity two- to threefold in rat liver, kidney, colon, cerebellum, and heart. Dose-response relationships revealed that at concentrations as low as 1 micromolar, both biotin and its analog caused maximal augmentation of guanylate cyclase activity. These data suggest a role for the activation of guanylate cyclase in the mechanism of action of this vitamin.
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.