Merck
  • Identification of intramolecular crosslinks in bovine growth hormone after two-step modification with 1,5-difluoro-2,4-dinitrobenzene.
Identification of intramolecular crosslinks in bovine growth hormone after two-step modification with 1,5-difluoro-2,4-dinitrobenzene.
International journal of peptide and protein research (1987-09-01)
M R Ermácora, C Nowicki, C Wolfenstein-Todel, J A Santomé
ABSTRACT
Derivatives of bovine growth hormone, containing monoaminotyrosyl residues in positions 35, 42 and 174, were treated at pH 3.6 with a bifunctional reagent, 1,5-difluoro-2,4-dinitrobenzene. Under these conditions aminotyrosyl groups reacted. On changing the pH to 9.3, the second fluorine atom of the reagent was substituted with the sterically adjacent side groups of lysine, since the excess of reagent had been previously removed. The modified protein underwent cyanogen bromide treatment. Peptides containing the crosslinks were purified from tryptic digests of the cyanogen bromide fragments by HPLC. Results show that aminoTyr 174 was able to form dinitrophenylene bridges with Lys 111, Lys 29 and Lys 170. AminoTry 35 was found crosslinked to Lys 29. Taking into account the size of the reagent, it may be inferred that Lys 29, 111 and 170 are located at approximately 5 A from Tyr 174 in the bovine growth hormone molecule.
Sigma-Aldrich
1,5-Difluoro-2,4-dinitrobenzene, 97%