Merck
  • [Chemical constituents from ethyl acetate extract of Shiaria bambusicola].
[Chemical constituents from ethyl acetate extract of Shiaria bambusicola].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-07-16)
Zhi-Qi Yin, Zhan-Li Chen, Jian Zhang, Wen-Cai Ye, Shou-Xun Zhao
ABSTRACT
Fourteen compounds were isolated and purified from the ethyl acetate of the ethanol extract of Shiaria bambusicola by various chromatographic methods, and their structures were elucidated by spectral techniques and physicochemical properties as hypocrellin A (1), hypocrellin B (2), hypocrellin C (3), hypomycin A (4), ergosterol (5), ergosterol peroxide (6), (22E, 24R)-5alpha, 8alpha-epidioxy-6,9(11),22-trien-3beta-ol (7), ergosta-7, 24(28)-dien-3beta-ol (8), (22E, 24R)-ergost-7, 22-dien-3beta, 5alpha, 6beta-triol (9), (22E,24R)-ergosta-7, 22-diene-3beta, 5alpha, 6beta-triol-3-O-palmitate (10), (22E, 24R)-ergosta-7, 22-diene-3beta, 5alpha, 6beta-triol-6-O-palmitate (11), 1-O-monostearin (12), 1, 3-O-distearin (13), and mannitol (14). Among them, compounds 7-13 were firstly isolated from this genus.
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Millipore
Ethyl acetate, suitable for HPLC, ≥99.8% (GC)
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%