Merck
[Progress with management of hereditary angioedema].
MMW Fortschritte der Medizin (2013-05-18)
D T Johnston, H Lode
ABSTRACT
Hereditary angioedema (HAE) is a rare type of angioedema caused by a quantitative or functional deficit of C1 inhibitor (C1 INH) that leads to excess production of bradykinin, which can result in acute localized swelling attacks in the skin or mucous membranes of the mouth, head and neck, extremities, gastrointestinal (GI) tract, genitals, trunk, and larynx. Angioedema in the respiratorytract maycause airway obstruction; severe abdominal pain, vomiting, or diarrhea may occur in the GI tract. Patients with HAE may be diagnosed and managed by HAE specialists or by primary care physicians depending on individual circumstances. Proper treatment requires differentiation from other forms of angioedema. Patients with HAE who are managed appropriately with medications that treat and prevent atttacks may have a lower risk of death from laryngeal edema and a better quality of life. Less frequent attacks may allow them to attend work, school, and leisure activities more regularlyand be free of the pain and disfigurement of HAE attacks moreoften.
Sigma-Aldrich
Bradykinin acetate salt, powder, ≥98% (HPLC)
Sigma-Aldrich
HOE 140, ≥94%