Merck
  • Two novel nanoscale preparations of micelle and thermosensitive hydrogel for docetaxel to treat malignant tumor.
Two novel nanoscale preparations of micelle and thermosensitive hydrogel for docetaxel to treat malignant tumor.
Journal of biomedical nanotechnology (2013-04-30)
Cheng Wang, Chaofeng Long, Chengshi Xie, Xiaoxin Chen, Lan Zhang, Bingyang Chu, Yujun Wang, Feng Luo, Zhiyong Qian
ABSTRACT
In this paper, two nanoscale preparations were described for docetaxel encapsulation using poly(epsilon-caprolactone)poly(ethylene glycol)-poly(epsilon-caprolactone) (PCEC) copolymer as carrier for treating malignant tumor. The first formulation was docetaxel-loaded PCEC micelle (D-M), which was characterized by XRD, TEM and Malvern laser particle size and drug release studies. The highest drug-loading of docetaxel in micelle was about 22.1 +/- 1.9%, optimized average diameter and polydispersity index was 25.2 +/- 1.1 nm, 0.13 +/- 0.12, respectively. Another formulation was docetaxel-loaded PCEC thermosensitive hydrogel (D-H), which displayed special gel-sol transition behavior with body temperature. We studied the cytotoxicity and in vitro hemolytic test of blank PCEC copolymer, the result was superiority. The data of relative body weight (RW), relative tumor volume (RV) and micrographs of hematoxylin and eosin (H&E)-stained histological sections showed D-M and D-H had significant antitumor effect and exhibited different characteristics of antitumor activity. Thus, the experiments signified that the combination therapy of intravenous (i.v.) and intratumoral administration using the two formulations maybe an effective way to treat malignant tumor.
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor