Merck
  • Statistical considerations when using a composite endpoint for comparing treatment groups.
Statistical considerations when using a composite endpoint for comparing treatment groups.
Statistics in medicine (2012-08-03)
Guadalupe Gómez, Stephen W Lagakos
ABSTRACT
When comparing two treatment groups in a time-to-event analysis, it is common to use a composite event consisting of two or more distinct outcomes. The goal of this paper is to develop a statistical methodology to derive efficiency guidelines for deciding whether to expand a study primary endpoint from E1 (for example, non-fatal myocardial infarction and cardiovascular death) to the composite of E1 and E2 (for example, non-fatal myocardial infarction, cardiovascular death or revascularisation). We investigate this problem by considering the asymptotic relative efficiency of a log-rank test for comparing treatment groups with respect to a primary relevant endpoint E1 versus the composite primary endpoint, say E, of E1 and E2, where E2 is some additional endpoint.
Product Description
Supelco
Probucol, analytical standard
Pricing and availability is not currently available.