Merck
  • Ionic liquid-nanoparticle hybrid electrolytes and their application in secondary lithium-metal batteries.
Ionic liquid-nanoparticle hybrid electrolytes and their application in secondary lithium-metal batteries.
Advanced materials (Deerfield Beach, Fla.) (2012-07-13)
Yingying Lu, Shyamal K Das, Surya S Moganty, Lynden A Archer
ABSTRACT
Ionic liquid-tethered nanoparticle hybrid electrolytes comprised of silica nanoparticles densely grafted with imidazolium-based ionic liquid chains are shown to retard lithium dendrite growth in rechargeable batteries with metallic lithium anodes. The electrolytes are demonstrated in full cell studies using both high-energy Li/MoS(2) and high-power Li/TiO(2) secondary batteries.
Sigma-Aldrich
Propylene carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Propylene carbonate, anhydrous, 99.7%
Sigma-Aldrich
Propylene carbonate, suitable for HPLC, 99.7%