Merck
  • Voltammetric investigation of DNA damage induced by nitrofurazone and short-lived nitro-radicals with the use of an electrochemical DNA biosensor.
Voltammetric investigation of DNA damage induced by nitrofurazone and short-lived nitro-radicals with the use of an electrochemical DNA biosensor.
Biosensors & bioelectronics (2012-06-22)
Yongnian Ni, Pingping Wang, Serge Kokot
ABSTRACT
Electrochemical behavior of nitrofurazone (NFZ) was investigated with the use of cyclic voltammetry (CV) and differential pulse voltammetry (DPV) methods. The pH-dependence of NFZ was studied at a glassy carbon electrode (GCE) in ethanol/Britton-Robinson buffer (30:70), and short-lived nitro-radicals were generated by the reduction of NFZ at high pHs (>7.0). In the presence of DNA, the DPV peak current of NFZ decreased and the peak potential shifted negatively, which indicated that there was an electrostatic interaction between NFZ and DNA. An electrochemical dsDNA/GCE biosensor was prepared to study the DNA damage produced in the presence NFZ; this process was followed with the use of the Co(phen)(3)(2+) electroactive probe. Also, the oxidation peaks of guanosine (750 mV) and adenosine (980 mV) indicated that DNA damage was related directly to the nitro-radicals. Experiments demonstrated that DNA damage occurred via two different steps while NFZ was metabolized and nitro-radicals were produced. Novel work with AFM on the NFZ/DNA interaction supported the suggestion that in vivo, the nitro-radicals were more cytotoxic than the NFZ molecules. A linear DPV calibration plot was obtained for NFZ analysis at a modified dsDNA/GCE (concentration range: 2.50 × 10(-6)-3.75 × 10(-5) mol L(-1); limit of detection: 8.0 × 10(-7) mol L(-1)), and NFZ was determined successfully in pharmaceutical samples.
Product Description
Supelco
Nitrofurazone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Nitro-2-furaldehyde semicarbazone, ≥97.0% (HPLC)